2-Chloro-3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%

Supplier: Ambeed
Warning

A145482-100MG A145482-1G A145482-250MG
77259-000EA 43.26 USD
77259-000 77259-002 77259-004
2-Chloro-3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
2-Chloro-3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₃
MW: 268.54 g/mol
MDL Number: MFCD31580204
CAS Number: 1799612-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR