3,3-Dimethyl-3,4-dihydroquinoxalin-2(1H)-one 98%

Supplier: Ambeed
Warning

A145399-100MG A145399-1G A145399-250MG
77235-172EA 22.66 USD
77235-172 77235-174 77235-176
3,3-Dimethyl-3,4-dihydroquinoxalin-2(1H)-one 98%
3,3-Dimethyl-3,4-dihydroquinoxalin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
MDL Number: MFCD00186947
CAS Number: 80636-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR