trans-ISRIB 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-trans-1,4-cyclohexanediylbis[2-(4-chlorophenoxy)acetamide], ISRIB, ISRIB (trans-isomer), N,N'-(trans-cyclohexane-1,4-diyl)bis(2-(4-chlorophenoxy)acetamide)

A145330-10MG A145330-1MG A145330-50MG A145330-5MG A145330-1G A145330-100MG
77235-158EA 48 USD
77235-158 77235-160 77235-162 77235-164 77566-936 77615-162
trans-ISRIB 98%
trans-ISRIB

Precaución: For research use only.

Formula: C₂₂H₂₄Cl₂N2₂O₄
MW: 451.3 g/mol
MDL Number: MFCD27952932
CAS Number: 1597403-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR