Ethyl 5-bromo-2-(trifluoromethyl)isonicotinate 95%

Supplier: Ambeed
Warning

A145251-100MG A145251-1G A145251-250MG
77258-994EA 21.63 USD
77258-994 77258-996 77258-998
Ethyl 5-bromo-2-(trifluoromethyl)isonicotinate 95%
Ethyl 5-bromo-2-(trifluoromethyl)isonicotinate

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO₂
MW: 298.06 g/mol
MDL Number: MFCD28746904
CAS Number: 1805222-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR