2-Bromo-5-methyl-1,3,4-thiadiazole 95%

Supplier: Ambeed
Warning

A145131-1G A145131-25G A145131-5G A145131-250MG
77285-898EA 10.3 USD
77285-898 77285-900 77285-902 77748-012
2-Bromo-5-methyl-1,3,4-thiadiazole 95%
2-Bromo-5-methyl-1,3,4-thiadiazole

Precaución: for Research Use Only

Formula: C₃H₃BrN₂S
MW: 179.04 g/mol
MDL Number: MFCD00518965
CAS Number: 54044-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR