4-(2-Aminopyrimidin-5-yl)benzoic acid 98%

Supplier: Ambeed
Warning

A144910-100MG A144910-1G A144910-250MG
77285-892EA 17.51 USD
77285-892 77285-894 77285-896
4-(2-Aminopyrimidin-5-yl)benzoic acid 98%
4-(2-Aminopyrimidin-5-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₉N₃O₂
MW: 215.2100 g/mol
MDL Number: MFCD05864814
CAS Number: 222987-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR