(1R,3S,4S)-tert-Butyl 3-(6-(7-(2-((S)-5-(tert-butoxycarbonyl)-5-azaspiro[2.4]heptan-6-yl)-1H-imidazol-5-yl)-9,9-difluoro-9H-fluoren-2-yl)-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate oxalate 97%

Supplier: Ambeed
Warning

A144838-100MG A144838-250MG
77258-972EA 23.69 USD
77258-972 77258-974
(1R,3S,4S)-tert-Butyl 3-(6-(7-(2-((S)-5-(tert-butoxycarbonyl)-5-azaspiro[2.4]heptan-6-yl)-1H-imidazol-5-yl)-9,9-difluoro-9H-fluoren-2-yl)-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate oxalate 97%
(1R,3S,4S)-tert-Butyl 3-(6-(7-(2-((S)-5-(tert-butoxycarbonyl)-5-azaspiro[2.4]heptan-6-yl)-1H-imidazol-5-yl)-9,9-difluoro-9H-fluoren-2-yl)-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate oxalate

Precaución: for Research Use Only

Formula: C₄₇H₅₀F₂N₆O₈
MW: 864.93 g/mol
MDL Number: MFCD30730079
CAS Number: 1499193-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR