3-Amino-4-methylpyridazine 95%

Supplier: Ambeed
Warning

A144829-100MG A144829-250MG
77272-666EA 96.82 USD
77272-666 77272-668
3-Amino-4-methylpyridazine 95%
3-Amino-4-methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.1300 g/mol
MDL Number: MFCD08460975
CAS Number: 90568-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR