4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetrabenzoic acid 95%

Supplier: Ambeed
Warning

A144745-100MG A144745-1G A144745-250MG A144745-5G
77258-964EA 29.87 USD
77258-964 77258-966 77258-968 77258-970
4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetrabenzoic acid 95%
4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetrabenzoic acid

Precaución: for Research Use Only

Formula: C₃₀H₂₀O₈
MW: 508.47 g/mol
MDL Number: MFCD27978395
CAS Number: 1351279-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR