2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinemethanol 97%

Supplier: Ambeed
Warning

A144722-100G A144722-10G A144722-1G A144722-250MG A144722-25G
77272-650EA 154 USD
77272-650 77272-652 77272-654 77272-656 77272-658
2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinemethanol 97%
2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinemethanol

Precaución: for Research Use Only

Formula: C₁₉H₁₆FNO
MDL Number: MFCD09032924
CAS Number: 121660-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR