4-Amino-5,6-dichlorobenzene-1,3-disulfonamide 97%

Supplier: Ambeed
Warning

A144712-100MG A144712-250MG A144712-1G
77285-884EA 59.74 USD
77285-884 77285-886 77748-011
4-Amino-5,6-dichlorobenzene-1,3-disulfonamide 97%
4-Amino-5,6-dichlorobenzene-1,3-disulphonamide

Precaución: for Research Use Only

Formula: C₆H₇Cl₂N₃O₄S₂
MW: 320.1735 g/mol
MDL Number: MFCD00111296
CAS Number: 5250-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR