N-Carbobenzoxy-DL-alanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cbz-DL-Ala-OH, N-Cbz-DL-alanine, 2-(((Benzyloxy)carbonyl)amino)propanoic acid, Z-DL-Ala-OH
A144621-25G
A144621-5G
77272-646EA
14
USD
77272-646
77748-008
N-Carbobenzoxy-DL-alanine 98%
N-Carbobenzoxy-DL-alanine
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol |
MDL Number:
MFCD00063125 CAS Number: 4132-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...