Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-3,5-diaminopyrazine-2-carboxylate, Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate

A144498-100G A144498-10G A144498-1G A144498-25G A144498-5G
77272-636EA 151.41 USD
77272-636 77272-638 77272-640 77272-642 77272-644
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate 98%
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇ClN₄O₂
MW: 202.60 g/mol
MDL Number: MFCD01928388
CAS Number: 1458-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR