N-Ethylpyridine-4-methylamine 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Pyridylmethyl)ethylamine, 4-(Ethylaminomethyl)pyridine

A144389-100G A144389-25G A144389-5G
77285-868EA 248 USD
77285-868 77285-870 77285-872
N-Ethylpyridine-4-methylamine 98%
N-Ethylpyridine-4-methylamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD00023632
CAS Number: 33403-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR