(R)-2-Amino-2-(naphthalen-1-yl)acetic acid hydrochloride 97%

Supplier: Ambeed
Warning

A144264-100MG A144264-250MG A144264-1G
77258-954EA 52.53 USD
77258-954 77258-956 77748-006
(R)-2-Amino-2-(naphthalen-1-yl)acetic acid hydrochloride 97%
(R)-2-Amino-2-(naphthalen-1-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClNO₂
MW: 237.68 g/mol
MDL Number: MFCD30749197
CAS Number: 1192350-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR