3-(4-Methylpiperidine-1-carbonyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-(4-Methylpiperidine-1-carbonyl)phenyl)boronic acid

A144225-1G A144225-250MG A144225-5G
77272-616EA 53.56 USD
77272-616 77272-618 77272-620
3-(4-Methylpiperidine-1-carbonyl)phenylboronic acid 95%
3-(4-Methylpiperidine-1-carbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₃
MW: 247.0979 g/mol
MDL Number: MFCD04115704
CAS Number: 850567-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR