2-Chloro-5-(methylthio)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Carboxy-4-chlorothioanisole, 2-Chloro-5-(methylsulphanyl)benzoic acid
A144104-10G
A144104-1G
A144104-250MG
A144104-5G
A144104-25G
77258-946EA
107
USD
77258-946
77258-948
77258-950
77258-952
77748-005
2-Chloro-5-(methylthio)benzoic acid 97%
2-Chloro-5-(methylthio)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇ClO₂S |
MDL Number:
MFCD00009721 CAS Number: 51546-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...