6-Bromo-2-hydroxyquinolin-4(1H)-one 95%

Supplier: Ambeed
Warning

A144086-1G A144086-250MG
77258-942EA 40.17 USD
77258-942 77258-944
6-Bromo-2-hydroxyquinolin-4(1H)-one 95%
6-Bromo-2-hydroxyquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.05 g/mol
MDL Number: MFCD29472381
CAS Number: 2254506-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR