2-(Ethylthio)pyrimidine-4,5,6-triamine 97%

Supplier: Ambeed
Warning

A143923-100MG A143923-1G A143923-250MG A143923-5G
77258-930EA 8.24 USD
77258-930 77258-932 77258-934 77258-936
2-(Ethylthio)pyrimidine-4,5,6-triamine 97%
2-(Ethylthio)pyrimidine-4,5,6-triamine

Precaución: for Research Use Only

Formula: C₆H₁₁N₅S
MW: 185.25 g/mol
MDL Number: MFCD20697420
CAS Number: 73000-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR