3,5-Difluorobenzophenone 95%

Supplier: Ambeed
Warning

A143911-10G A143911-25G A143911-5G
77272-602EA 22 USD
77272-602 77272-604 77272-606
3,5-Difluorobenzophenone 95%
3,5-Difluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈F₂O
MDL Number: MFCD00061140
CAS Number: 179113-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR