4-Chloro-3-methoxy-2-methylpyridine 97%

Supplier: Ambeed
Warning

A143864-100MG A143864-1G A143864-250MG A143864-5G
77285-852EA 21 USD
77285-852 77285-854 77285-856 77615-148
4-Chloro-3-methoxy-2-methylpyridine 97%
4-Chloro-3-methoxy-2-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MDL Number: MFCD16610605
CAS Number: 107512-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR