3,5-Diamino-4-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A143854-1G A143854-250MG A143854-5G
77272-596EA 20 USD
77272-596 77272-598 77272-600
3,5-Diamino-4-methylbenzoic acid 97%
3,5-Diamino-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00025283
CAS Number: 6633-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR