1H,1H,2H,2H-Perfluorooctyltriethoxysilane 96%

Supplier: Ambeed
Warning

Synonyms: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;

A143780-100G A143780-10G A143780-1G A143780-25G A143780-5G
77235-116EA 129.78 USD
77235-116 77235-118 77235-120 77235-122 77235-124
1H,1H,2H,2H-Perfluorooctyltriethoxysilane 96%
1H,1H,2H,2H-Perfluorooctyltriethoxysilane

Precaución: for Research Use Only

Formula: C₁₄H₁₉F₁₃O₃Si
MW: 510.367 g/mol
MDL Number: MFCD00042333
CAS Number: 51851-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR