N-Boc-L-threonine 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-threonine, Boc-Thr-OH, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid

A143765-10G A143765-500G A143765-5G A143765-100G A143765-25G A143765-1000G
77272-588EA 9.27 USD
77272-588 77272-590 77272-592 77566-884 77566-886 77748-004
N-Boc-L-threonine 95%
N-Boc-L-threonine

Precaución: For research use only.

Formula: C₉H₁₇NO₅
MW: 219.2 g/mol
MDL Number: MFCD00065946
CAS Number: 2592-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR