5-Bromo-2,4-di(tert-butoxy)pyrimidine 95%

Supplier: Ambeed
Warning

A143634-100MG A143634-1G A143634-250MG A143634-5G
77247-060EA 14 USD
77247-060 77247-062 77247-064 77247-066
5-Bromo-2,4-di(tert-butoxy)pyrimidine 95%
5-Bromo-2,4-di(tert-butoxy)pyrimidine

Precaución: for Research Use Only

Formula: C₁₂H₁₉BrN₂O₂
MDL Number: MFCD01927527
CAS Number: 19752-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR