(E)-Methyl 3-(methoxy(phenyl)methylene)-2-oxoindoline-6-carboxylate 97%

Supplier: Ambeed
Warning

A143608-100MG A143608-1G A143608-250MG A143608-25G A143608-5G
77235-104EA 8 USD
77235-104 77235-106 77235-108 77235-110 77235-112
(E)-Methyl 3-(methoxy(phenyl)methylene)-2-oxoindoline-6-carboxylate 97%
(E)-Methyl 3-(methoxy(phenyl)methylene)-2-oxoindoline-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₄
MDL Number: MFCD28139165
CAS Number: 1168150-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR