3-(3-((Trifluoromethyl)thio)phenyl)-1H-pyrazol-5-amine 97%

Supplier: Ambeed
Warning

A143601-1G A143601-250MG
77247-054EA 27.81 USD
77247-054 77247-056 77247-058
3-(3-((Trifluoromethyl)thio)phenyl)-1H-pyrazol-5-amine 97%
3-(3-((Trifluoromethyl)thio)phenyl)-1H-pyrazol-5-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₃N₃S
MDL Number: MFCD24539382
CAS Number: 1912400-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR