(16aR)-1,16-Bis(diphenylphosphino)-6,7,8,9,10,11-hexahydrodibenzo[b,d][1,6]dioxacyclododecine 98%

Supplier: Ambeed
Warning

A143549-100MG A143549-250MG
77258-926EA 110 USD
77258-926 77258-928
(16aR)-1,16-Bis(diphenylphosphino)-6,7,8,9,10,11-hexahydrodibenzo[b,d][1,6]dioxacyclododecine 98%
(16aR)-1,16-Bis(diphenylphosphino)-6,7,8,9,10,11-hexahydrodibenzo[b,d][1,6]dioxacyclododecine

Precaución: for Research Use Only

Formula: C₄₂H₃₈O₂P₂
CAS Number: 301847-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR