2-Hydroxyquinoline-3-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2(1H)-Quinolinone-3-carboxaldehyde, 2-Oxo-1,2-dihydro-3-quinolinecarbaldehyde

A143538-1G A143538-250MG A143538-5G
77272-580EA 88.58 USD
77272-580 77272-582 77748-003
2-Hydroxyquinoline-3-carbaldehyde 95%
2-Hydroxyquinoline-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.171 g/mol
MDL Number: MFCD00563443
CAS Number: 91301-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR