(R)-2-Amino-3-((2-boronoethyl)thio)propanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: BEC·HCl

A143511-10MG A143511-1MG A143511-25MG A143511-50MG A143511-5MG
77258-916EA 120.51 USD
77258-916 77258-918 77258-920 77258-922 77258-924
(R)-2-Amino-3-((2-boronoethyl)thio)propanoic acid hydrochloride 97%
(R)-2-Amino-3-((2-boronoethyl)thio)propanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₃BClNO₄S
MW: 229.49 g/mol
MDL Number: MFCD03453539
CAS Number: 222638-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR