4-Methylpyridazine 97%

Supplier: Ambeed
Warning

A143474-1G A143474-5G
77285-848EA 16.48 USD
77285-848 77285-850
4-Methylpyridazine 97%
4-Methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₆N₂
MW: 94.12 g/mol
MDL Number: MFCD00011594
CAS Number: 1120-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR