(E)-6-Iodo-3-(2-(pyridin-2-yl)vinyl)-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole 98%

Supplier: Ambeed
Warning

A143415-10G A143415-1G A143415-250MG A143415-5G
77247-034EA 355.99 USD
77247-034 77247-036 77247-038 77247-040
(E)-6-Iodo-3-(2-(pyridin-2-yl)vinyl)-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole 98%
(E)-6-Iodo-3-(2-(pyridin-2-yl)vinyl)-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₉H₁₈IN₃O
MDL Number: MFCD12405576
CAS Number: 886230-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR