2-(3-Chlorophenyl)acetamide 98%

Supplier: Ambeed
Warning

A143372-1G A143372-250MG
77235-092EA 78.28 USD
77235-092 77235-094
2-(3-Chlorophenyl)acetamide 98%
2-(3-Chlorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.608 g/mol
MDL Number: MFCD09991645
CAS Number: 58357-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR