4-Bromo-2-methylthiophene 98%

Supplier: Ambeed
Warning

A143360-100G A143360-10G A143360-1G A143360-25G A143360-5G
77285-838EA 147 USD
77285-838 77285-840 77285-842 77285-844 77285-846
4-Bromo-2-methylthiophene 98%
4-Bromo-2-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₅BrS
MDL Number: MFCD04116367
CAS Number: 29421-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR