(S)-2-Amino-2-(3-fluorophenyl)ethanol hydrochloride 97%

Supplier: Ambeed
Warning

A143270-1G A143270-250MG
77247-030EA 220.42 USD
77247-030 77247-032
(S)-2-Amino-2-(3-fluorophenyl)ethanol hydrochloride 97%
(S)-2-Amino-2-(3-fluorophenyl)ethanol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClFNO
MW: 191.63 g/mol
MDL Number: MFCD12758113
CAS Number: 1240480-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR