2-(4-Nitrophenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A143269-1G A143269-250MG A143269-5G
77272-572EA 10 USD
77272-572 77272-574 77272-576
2-(4-Nitrophenyl)propanoic acid 95%
2-(4-Nitrophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.17 g/mol
MDL Number: MFCD00012106
CAS Number: 19910-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR