2-(3-Chlorophenyl)-4,6-diphenyl-1,3,5-triazine 98%

Supplier: Ambeed
Warning

A143186-1G A143186-25G A143186-5G
77235-080EA 6.18 USD
77235-080 77235-082 77235-084
2-(3-Chlorophenyl)-4,6-diphenyl-1,3,5-triazine 98%
2-(3-Chlorophenyl)-4,6-diphenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₄ClN₃
MW: 343.80 g/mol
MDL Number: MFCD25562934
CAS Number: 307929-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR