3-(2-Aminoethyl)thiophene hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Thiophen-3-yl)ethanamine hydrochloride, 2-(Thien-3-yl)ethylamine hydrochloride

A143166-1G A143166-250MG A143166-5G
77272-566EA 55.62 USD
77272-566 77272-568 77272-570
3-(2-Aminoethyl)thiophene hydrochloride 97%
3-(2-Aminoethyl)thiophene hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉NS·ClH
MW: 163.668 g/mol
MDL Number: MFCD09265521
CAS Number: 34843-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR