MCB613 98%

Supplier: Ambeed
Warning

Synonyms: 4-Ethyl-2,6-bis(pyridin-3-ylmethylene)cyclohexanone, 4-Ethyl-2,6-bis(3-pyridinylmethylene)-cyclohexanone

A143145-25MG A143145-5MG
77235-060EA 938.33 USD
77235-060 77235-062 77235-064 77235-066 77235-068 77235-070
MCB613 98%
MCB613

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O
MDL Number: MFCD01209975
CAS Number: 1162656-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR