3-Amino-p-anisic acid 95%

Supplier: Ambeed
Warning

A143141-100G A143141-1G A143141-25G A143141-5G
77247-022EA 82 USD
77247-022 77247-024 77247-026 77247-028
3-Amino-p-anisic acid 95%
3-Amino-p-anisic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD00002521
CAS Number: 2840-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR