N-(2-Aminoethyl)maleimide hydrochloride 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(2-Aminoethyl)maleimide HCl, N-(2-Aminoethyl)maleimide Hydrochloride, 2-Maleimidoethylamine hydrochloride
A143117-100MG
A143117-1G
A143117-250MG
A143117-25G
A143117-5G
A143117-100G
77258-900EA
5
USD
77258-900
77258-902
77258-904
77258-906
77258-908
77748-002
N-(2-Aminoethyl)maleimide hydrochloride 96%
N-(2-Aminoethyl)maleimide hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 176.60 g/mol |
MDL Number:
MFCD09800491 CAS Number: 134272-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...