2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxymethyl-2-ethoxybenzoic Acid Ethyl Ester, Ethyl 4-Carboxymethyl-2-ethoxybenzoate, 2-(3-Ethoxy-4-(ethoxycarbonyl)phenyl)acetic acid

A142970-100G A142970-10G A142970-1G A142970-25G A142970-5G
77272-552EA 158 USD
77272-552 77272-554 77272-556 77272-558 77272-560
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid 98%
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₅
MW: 252.27 g/mol
MDL Number: MFCD08704233
CAS Number: 99469-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR