3-(4-Hydroxy-3-nitrophenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A142923-100MG A142923-250MG
77272-548EA 11 USD
77272-548 77272-550
3-(4-Hydroxy-3-nitrophenyl)propanoic acid 95%
3-(4-Hydroxy-3-nitrophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MW: 211.18 g/mol
MDL Number: MFCD00045947
CAS Number: 38196-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR