VR23 ≥98%

Supplier: Ambeed
Warning

Synonyms: 7-chloro-4-(4-((2,4-dinitrophenyl)sulphonyl)piperazin-1-yl)quinoline

A142790-1G A142790-10MG A142790-5MG
77235-058EA 468.65 USD
77235-058 77746-006 77746-007
VR23 ≥98%
VR23

Precaución: for Research Use Only

Formula: C₁₉H₁₆ClN₅O₆S
MW: 477.88 g/mol
MDL Number: MFCD28502093
CAS Number: 1624602-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR