2-Fluoro-5-(hydroxymethyl)benzeneboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-(hydroxymethyl)phenyl)boronic acid

A142781-10G A142781-1G A142781-25G
77272-542EA 49.44 USD
77272-542 77272-544 77272-546
2-Fluoro-5-(hydroxymethyl)benzeneboronic acid 96%
2-Fluoro-5-(hydroxymethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD06656276
CAS Number: 1072952-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR