2,4-Dibromo-6-nitroaniline 98+%

Supplier: Ambeed
Warning

A142624-100G A142624-1G A142624-25G A142624-5G
77247-002EA 478.95 USD
77247-002 77247-004 77247-006 77247-008
2,4-Dibromo-6-nitroaniline 98+%
2,4-Dibromo-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂N₂O₂
MW: 295.92 g/mol
MDL Number: MFCD00012107
CAS Number: 827-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR