Methyl 4-acetyl-2-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A142505-100MG A142505-250MG A142505-1G
77285-816EA 123 USD
77285-816 77285-818 77747-998
Methyl 4-acetyl-2-hydroxybenzoate 95%
Methyl 4-acetyl-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD09907817
CAS Number: 27475-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR