L(+)-Lactic acid 95%

Supplier: Ambeed
Warning

Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

A142479-100G A142479-25G A142479-500G
77258-876EA 32.96 USD
77258-876 77258-878 77258-880 77258-882
L(+)-Lactic acid 95%
L(+)-Lactic acid

Precaución: for Research Use Only

Formula: C₃H₆O₃
MDL Number: MFCD00064266
CAS Number: 79-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR