Ethyl-3-(4-fluorophenyl)-5-methylisoxazole-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(4-fluorophenyl)-5-methylisoxazole-4-carboxylate

A142413-1G A142413-250MG
77272-532EA 173.04 USD
77272-532 77272-534
Ethyl-3-(4-fluorophenyl)-5-methylisoxazole-4-carboxylate 98%
Ethyl-3-(4-fluorophenyl)-5-methylisoxazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₂FNO₃
MW: 249.24 g/mol
MDL Number: MFCD09756447
CAS Number: 954230-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR