(E)-3-(2-Propyl-6-(trifluoromethyl)pyridin-3-yl)acrylic acid 97%

Supplier: Ambeed
Warning

A142397-100MG A142397-1G A142397-250MG
77235-038EA 5 USD
77235-038 77235-040 77235-042
(E)-3-(2-Propyl-6-(trifluoromethyl)pyridin-3-yl)acrylic acid 97%
(E)-3-(2-Propyl-6-(trifluoromethyl)pyridin-3-yl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₃NO₂
MDL Number: MFCD30478615
CAS Number: 1005174-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR